COM448681404 Combi-Blocks, Inc.

6-Acetylamino-2-spiro(n-boc-piperidine-4-yl)-benzopyran

  • MDL number MFCD07369808
  • Cas Number : 886363-78-6
  • Molecular Weight : 360.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5377
Availability details for
Quantity
  • Availability
  • Add to Cart