COM448681406 Combi-Blocks, Inc.

6-Acetylamino-4-oxo-2-spiro(n-boc-piperidine-4-yl)-benzopyran

  • MDL number MFCD07369807
  • Cas Number : 223559-44-2
  • Molecular Weight : 374.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5379

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5379
Availability details for
Quantity
  • Availability
  • Add to Cart