COM448681441 Combi-Blocks, Inc.

6-Aminomethyl-spiro[2.5]octane

  • MDL number MFCD11501146
  • Cas Number : 877201-35-9
  • Molecular Weight : 139.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5435
Availability details for
Quantity
  • Availability
  • Add to Cart