COM448681472 Combi-Blocks, Inc.

6-Bromo-1,2,3,4-tetrahydro-quinoline-2-carboxylic acid methyl ester

  • MDL number MFCD03426372
  • Cas Number : 511230-72-1
  • Molecular Weight : 270.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5513
Availability details for
Quantity
  • Availability
  • Add to Cart