COM448681492 Combi-Blocks, Inc.

6-Bromo-2',3',5',6'-tetrahydrospiro[indene-2,4'-pyran]-1(3h)-one

  • MDL number MFCD18711529
  • Cas Number : 1245514-78-6
  • Molecular Weight : 281.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5556

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5556
Availability details for
Quantity
  • Availability
  • Add to Cart