COM448681494 Combi-Blocks, Inc.

6-Bromo-2,3-dimethylimidazo[1,2-a]pyrimidine

  • MDL number MFCD12024555
  • Cas Number : 1111638-07-3
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5561
Availability details for
Quantity
  • Availability
  • Add to Cart