COM448681631 Combi-Blocks, Inc.

6-Bromo-8-(methylthio)imidazo[1,2-a]pyrazine

  • MDL number MFCD11100003
  • Cas Number : 887475-71-0
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5764
Availability details for
Quantity
  • Availability
  • Add to Cart