COM448681668 Combi-Blocks, Inc.

6-Bromo-quinoline-2-carbonitrile

  • MDL number MFCD08062622
  • Cas Number : 65185-41-3
  • Molecular Weight : 233.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5810

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5810
Availability details for
Quantity
  • Availability
  • Add to Cart