COM448681701 Combi-Blocks, Inc.

6-Chloro-2-methyl-thiazolo[5,4-b]pyridine

  • MDL number MFCD07375042
  • Cas Number : 857970-22-0
  • Molecular Weight : 184.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5862
Availability details for
Quantity
  • Availability
  • Add to Cart