COM448681714 Combi-Blocks, Inc.

6-Chloro-4-methyl-7-azaindole

  • MDL number MFCD08061194
  • Cas Number : 4894-29-5
  • Molecular Weight : 166.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5879
Availability details for
Quantity
  • Availability
  • Add to Cart