COM448681734 Combi-Blocks, Inc.

3-(Boc-amino)-6-fluoro-1h-indazole

  • MDL number MFCD18651681
  • Cas Number : 1176089-41-0
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5910

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5910
Availability details for
Quantity
  • Availability
  • Add to Cart