COM448681786 Combi-Blocks, Inc.

6-Chloro-trans-2-hexenoic acid ethyl ester

  • MDL number MFCD05863887
  • Cas Number : 72448-92-1
  • Molecular Weight : 176.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5986
Availability details for
Quantity
  • Availability
  • Add to Cart