COM448681796 Combi-Blocks, Inc.

6-Cyclohexyl-3(2h)-pyridazinone

  • MDL number MFCD11520598
  • Cas Number : 105537-90-4
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6000
Availability details for
Quantity
  • Availability
  • Add to Cart