COM448681821 Combi-Blocks, Inc.

6-Fluoro-4-hydroxy-3-nitroquinoline

  • MDL number MFCD09800466
  • Cas Number : 628284-75-3
  • Molecular Weight : 208.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6044
Availability details for
Quantity
  • Availability
  • Add to Cart