COM448681836 Combi-Blocks, Inc.

6-Fmoc-hydrazinonicotinic acid

  • MDL number MFCD03788695
  • Cas Number : 444794-69-8
  • Molecular Weight : 375.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6067
Availability details for
Quantity
  • Availability
  • Add to Cart