COM448681849 Combi-Blocks, Inc.

6-Hydroxyspiro[chroman-2,4'-piperidin]-4-one, HCl

  • MDL number MFCD09998259
  • Cas Number : 1189985-17-8
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6086
Availability details for
Quantity
  • Availability
  • Add to Cart