COM448681992 Combi-Blocks, Inc.

6-N-Boc-aminoindole-4-carboxylic acid

  • MDL number MFCD09800410
  • Cas Number : 948015-63-2
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6324
Availability details for
Quantity
  • Availability
  • Add to Cart