COM448682014 Combi-Blocks, Inc.

6-tert-Butyl-pyridine-3-carbaldehyde

  • MDL number MFCD09029652
  • Cas Number : 391900-69-9
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6367
Availability details for
Quantity
  • Availability
  • Add to Cart