COM448682015 Combi-Blocks, Inc.

6-Tritylamino-nicotinic acid

  • MDL number MFCD03839996
  • Cas Number : 49647-10-1
  • Molecular Weight : 380.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6368
Availability details for
Quantity
  • Availability
  • Add to Cart