COM448682040 Combi-Blocks, Inc.

7-Amino-1h-benzo[d][1,3]oxazine-2,4-dione

  • MDL number MFCD03839875
  • Cas Number : 179331-04-5
  • Molecular Weight : 178.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6416

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6416
Availability details for
Quantity
  • Availability
  • Add to Cart