COM448682078 Combi-Blocks, Inc.

2-Chloro-6-fluoroquinazolin-4-amine

  • MDL number MFCD10697892
  • Cas Number : 192323-44-7
  • Molecular Weight : 197.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6475

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6475
Availability details for
Quantity
  • Availability
  • Add to Cart