COM448682105 Combi-Blocks, Inc.

7-Bromo-5-fluoro-3-methyl-1h-indole

  • MDL number MFCD14706303
  • Cas Number : 883001-24-9
  • Molecular Weight : 228.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6517
Availability details for
Quantity
  • Availability
  • Add to Cart