COM448682119 Combi-Blocks, Inc.

7-Chloro-3-hydroxy-3h-isobenzofuran-1-one

  • MDL number MFCD17926369
  • Cas Number : 70097-44-8
  • Molecular Weight : 184.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6544
Availability details for
Quantity
  • Availability
  • Add to Cart