COM448682124 Combi-Blocks, Inc.

7-Chlorocinnoline-3-carboxylic acid

  • MDL number MFCD17926281
  • Cas Number : 480451-01-2
  • Molecular Weight : 208.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6554
Availability details for
Quantity
  • Availability
  • Add to Cart