COM448682129 Combi-Blocks, Inc.

7-Chlorotryptamine, HCl

  • MDL number MFCD11501280
  • Cas Number : 81868-13-5
  • Molecular Weight : 231.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6561
Availability details for
Quantity
  • Availability
  • Add to Cart