COM448682139 Combi-Blocks, Inc.

7-Fluoro-2,3,4,5-tetrahydro-1h-benzo[e][1,4]diazepine

  • MDL number MFCD07369955
  • Cas Number : 886366-21-8
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6584
Availability details for
Quantity
  • Availability
  • Add to Cart