COM448682146 Combi-Blocks, Inc.

7-Formyl-8-quinolinol

  • MDL number MFCD09991697
  • Cas Number : 5683-78-3
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-hydroxy-7-quinolinecarbaldehyde
Inchi Code 1S/C10H7NO2/c12-6-8-4-3-7-2-1-5-11-9(7)10(8)13/h1-6,13H
Inchi Key HGNLFMRNEFPDHZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6597

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6597
Availability details for
Quantity
  • Availability
  • Add to Cart