COM448682169 Combi-Blocks, Inc.

7-Methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid

  • MDL number MFCD10687189
  • Cas Number : 24833-31-6
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6644

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6644
Availability details for
Quantity
  • Availability
  • Add to Cart