COM448682214 Combi-Blocks, Inc.

7-Methylthieno[3,2-d]pyrimidine

  • MDL number MFCD13181518
  • Cas Number : 871013-26-2
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6712

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6712
Availability details for
Quantity
  • Availability
  • Add to Cart