COM448682273 Combi-Blocks, Inc.

8-Benzyl-1-phenyl-1,3,8-triaza-spiro[4.5]decan-4-one

  • MDL number MFCD02929369
  • Cas Number : 974-41-4
  • Molecular Weight : 321.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6800

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6800
Availability details for
Quantity
  • Availability
  • Add to Cart