COM448682299 Combi-Blocks, Inc.

3-(Ethylaminosulfonyl)benzoic acid

  • MDL number MFCD06353541
  • Cas Number : 7326-74-1
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6842

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6842
Availability details for
Quantity
  • Availability
  • Add to Cart