COM448682318 Combi-Blocks, Inc.

6-Methoxy-1h-pyrido[2,3-b][1,4]oxazin-2(3h)-one

  • MDL number MFCD18910078
  • Cas Number : 1378342-69-8
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6883
Availability details for
Quantity
  • Availability
  • Add to Cart