COM448682324 Combi-Blocks, Inc.

Methyl 2-(5-oxazolyl)benzoate

  • MDL number MFCD18375278
  • Cas Number : 1261268-97-6
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6894

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6894
Availability details for
Quantity
  • Availability
  • Add to Cart