COM448682332 Combi-Blocks, Inc.

8-N-Boc-5,6,7,8-tetrahydro-1,8-naphthyridin-2-propionic acid

  • MDL number MFCD04115299
  • Cas Number : 886362-45-4
  • Molecular Weight : 306.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6907

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6907
Availability details for
Quantity
  • Availability
  • Add to Cart