COM448682347 Combi-Blocks, Inc.

8-O-Benzoyl-hydroxy-quinoline-7-carbaldehyde

  • MDL number MFCD09261304
  • Cas Number : 936497-79-9
  • Molecular Weight : 277.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6928

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6928
Availability details for
Quantity
  • Availability
  • Add to Cart