COM448682352 Combi-Blocks, Inc.

9,10-Bis(chloromethyl)anthracene

  • MDL number MFCD00045388
  • Cas Number : 10387-13-0
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6941

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6941
Availability details for
Quantity
  • Availability
  • Add to Cart