COM448682366 Combi-Blocks, Inc.

9-Chloro-2,3,4,5-tetrahydro-1h-benzo[e][1,4]diazepine 2hcl

  • MDL number MFCD11707062
  • Cas Number : 1159823-84-3
  • Molecular Weight : 255.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6971
Availability details for
Quantity
  • Availability
  • Add to Cart