COM448682367 Combi-Blocks, Inc.

9-Chloro-3,4-dihydro-1h-benzo[e][1,4]diazepine-2,5-dione

  • MDL number MFCD20261473
  • Cas Number : 1334499-55-6
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6973
Availability details for
Quantity
  • Availability
  • Add to Cart