COM448682377 Combi-Blocks, Inc.

9-Methyl-2,3,4,5-tetrahydro-1h-benzo[e][1,4]diazepine

  • MDL number MFCD07369976
  • Cas Number : 195986-82-4
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6992

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6992
Availability details for
Quantity
  • Availability
  • Add to Cart