COM448682390 Combi-Blocks, Inc.

Alpha-chloro-3-nitrobenzaldoxime

  • MDL number MFCD01924504
  • Cas Number : 33512-94-6
  • Molecular Weight : 200.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7009
Availability details for
Quantity
  • Availability
  • Add to Cart