COM448682405 Combi-Blocks, Inc.

N-Cbz-9-azabicyclo[3.3.1]nonan-3-one

  • MDL number MFCD11052330
  • Cas Number : 146747-65-1
  • Molecular Weight : 273.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7032

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7032
Availability details for
Quantity
  • Availability
  • Add to Cart