COM448682414 Combi-Blocks, Inc.

2-Chloro-1-(1-(methylsulfonyl)indolin-5-yl)ethanone

  • MDL number MFCD06378262
  • Cas Number : 851288-79-4
  • Molecular Weight : 273.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7048
Availability details for
Quantity
  • Availability
  • Add to Cart