COM448682455 Combi-Blocks, Inc.

Azepan-3-one, HCl

  • MDL number MFCD17926170
  • Cas Number : 65326-54-7
  • Molecular Weight : 149.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7106
Availability details for
Quantity
  • Availability
  • Add to Cart