COM448682456 Combi-Blocks, Inc.

Azepan-3-yl-methyl-amine

  • MDL number MFCD02683066
  • Cas Number : 124695-93-8
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7108
Availability details for
Quantity
  • Availability
  • Add to Cart