COM448682474 Combi-Blocks, Inc.

Benzhydrylidene methylamine

  • MDL number MFCD02179401
  • Cas Number : 13280-16-5
  • Molecular Weight : 195.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7141
Availability details for
Quantity
  • Availability
  • Add to Cart