COM448682501 Combi-Blocks, Inc.

Benzyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, HCl

  • MDL number MFCD00798224
  • Cas Number : 103733-30-8
  • Molecular Weight : 303.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7180
Availability details for
Quantity
  • Availability
  • Add to Cart