COM448682513 Combi-Blocks, Inc.

4-Benzyl-2-(hydroxymethyl)homomorpholine

  • MDL number MFCD14585013
  • Cas Number : 1031442-66-6
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7202

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7202
Availability details for
Quantity
  • Availability
  • Add to Cart