COM448682515 Combi-Blocks, Inc.

Bis(6-chloro-3-methylpyridin-2-yl)methanone

  • MDL number MFCD22887417
  • Cas Number : 1414864-03-1
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7207
Availability details for
Quantity
  • Availability
  • Add to Cart