COM448682549 Combi-Blocks, Inc.

Bis-(6-methyl-pyridin-2-yl)-amine

  • MDL number MFCD10687142
  • Cas Number : 85895-80-3
  • Molecular Weight : 199.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7266
Availability details for
Quantity
  • Availability
  • Add to Cart