COM448682561 Combi-Blocks, Inc.

Boc-(r)-3-amino-3-(3-pyridyl)-propionic acid

  • MDL number MFCD03427948
  • Cas Number : 500788-96-5
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7292
Availability details for
Quantity
  • Availability
  • Add to Cart